Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: leng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016263.2 | NP_001016263.1 | 1 - 2 | 164 | 65 | |
GenBank mRNA | BC170660.1 | - | 1 - 2 | 159 | 65 | |
BC170662.1 | - | 1 - 2 | 159 | 65 | ||
CR761695.2 | - | 1 - 2 | 164 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |