Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tdrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016270.2 | NP_001016270.1 | 3 - 4 | 411 | 103 | |
XM_012962978.2 | XP_012818432.1 | 4 - 5 | 833 | 103 | ||
GenBank mRNA | CR761669.2 | - | 3 - 4 | 411 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |