Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcyox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016302.2 | NP_001016302.1 | 2 - 3 | 334 | 94 | |
GenBank mRNA | BC170904.1 | - | 2 - 3 | 324 | 94 | |
BC170906.1 | - | 2 - 3 | 324 | 94 | ||
CR761521.2 | - | 2 - 3 | 334 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |