Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016327.2 | NP_001016327.1 | 7 - 8 | 990 | 93 | |
GenBank mRNA | BC136241.1 | - | 7 - 8 | 998 | 93 | |
BC155384.1 | - | 7 - 8 | 966 | 93 | ||
CR761429.2 | - | 7 - 8 | 990 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |