Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016333.2 | NP_001016333.1 | 3 - 4 | 1018 | 95 | |
XM_012962186.2 | XP_012817640.1 | 4 - 5 | 860 | 95 | ||
XM_012962189.2 | XP_012817643.1 | 4 - 5 | 859 | 95 | ||
XM_012962191.1 | XP_012817645.1 | 4 - 5 | 776 | 95 | ||
XM_018089476.1 | XP_017944965.1 | 3 - 4 | 526 | 95 | ||
GenBank mRNA | BC171112.1 | - | 3 - 4 | 930 | 95 | |
CR761413.2 | - | 3 - 4 | 1018 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |