Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016346.3 | NP_001016346.2 | 5 - 6 | 479 | 74 | |
XM_012956960.2 | XP_012812414.1 | 5 - 6 | 1552 | 74 | ||
XM_012956961.2 | XP_012812415.1 | 5 - 6 | 1610 | 74 | ||
GenBank mRNA | BC135519.1 | - | 5 - 6 | 466 | 74 | |
CR761368.2 | - | 4 - 5 | 436 | 74 | ||
CX821871.1 | - | 5 - 6 | 479 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |