Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016349.2 | NP_001016349.1 | 7 - 8 | 1018 | 101 | |
XM_012956962.1 | XP_012812416.1 | 7 - 8 | 1169 | 101 | ||
XM_012956963.2 | XP_012812417.1 | 7 - 8 | 1181 | 101 | ||
GenBank mRNA | BC167698.1 | - | 7 - 8 | 1149 | 101 | |
BC170828.1 | - | 7 - 8 | 1017 | 101 | ||
BC170830.1 | - | 7 - 8 | 1017 | 101 | ||
CR761344.2 | - | 7 - 8 | 1018 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |