Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pabpn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016352.2 | NP_001016352.1 | 1 - 2 | 140 | 63 | |
XM_012961085.2 | XP_012816539.1 | 2 - 3 | 459 | 63 | ||
XM_012961086.2 | XP_012816540.1 | 2 - 3 | 226 | 63 | ||
XM_012961087.2 | XP_012816541.1 | 2 - 3 | 245 | 63 | ||
GenBank mRNA | BC121546.1 | - | 2 - 3 | 351 | 63 | |
CR761333.2 | - | 1 - 2 | 140 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |