Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap4s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016360.2 | NP_001016360.1 | 1 - 2 | 176 | 92 | |
XM_012968396.2 | XP_012823850.1 | 2 - 3 | 247 | 92 | ||
XM_012968397.2 | XP_012823851.1 | 2 - 3 | 332 | 92 | ||
GenBank mRNA | BC135702.1 | - | 2 - 3 | 216 | 92 | |
CR761300.2 | - | 1 - 2 | 176 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |