Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016364.2 | NP_001016364.1 | 2 - 3 | 193 | 58 | |
GenBank mRNA | CR761293.2 | - | 2 - 3 | 193 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |