Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016384.2 | NP_001016384.1 | 4 - 5 | 1000 | 72 | |
GenBank mRNA | BC170897.1 | - | 4 - 5 | 976 | 72 | |
BC170901.1 | - | 4 - 5 | 976 | 72 | ||
CR761211.2 | - | 4 - 5 | 1000 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |