Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016397.2 | NP_001016397.1 | 1 - 2 | 206 | 71 | |
GenBank mRNA | BC170714.1 | - | 1 - 2 | 204 | 71 | |
BC170716.1 | - | 1 - 2 | 204 | 71 | ||
CR761158.2 | - | 1 - 2 | 206 | 71 | ||
CR761349.2 | - | 1 - 2 | 188 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |