Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp5f1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016407.2 | NP_001016407.1 | 5 - 6 | 639 | 67 | |
XM_012959013.1 | XP_012814467.1 | 5 - 6 | 722 | 67 | ||
XM_012959014.1 | XP_012814468.1 | 5 - 6 | 722 | 67 | ||
GenBank mRNA | CR761126.2 | - | 5 - 6 | 639 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons |