Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016422.2 | NP_001016422.1 | 4 - 5 | 596 | 86 | |
GenBank mRNA | BC170814.1 | - | 4 - 5 | 595 | 86 | |
BC170816.1 | - | 4 - 5 | 595 | 86 | ||
CR761072.2 | - | 4 - 5 | 596 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |