Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016424.2 | NP_001016424.1 | 4 - 5 | 293 | 88 | |
GenBank mRNA | CR761067.2 | - | 4 - 5 | 293 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |