Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016431.2 | NP_001016431.1 | 4 - 5 | 527 | 84 | |
GenBank mRNA | BC135174.2 | - | 4 - 5 | 460 | 84 | |
CR761051.2 | - | 4 - 5 | 527 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |