Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016433.2 | NP_001016433.1 | 4 - 5 | 1105 | 123 | |
XM_012968410.2 | XP_012823864.1 | 4 - 5 | 1271 | 123 | ||
GenBank mRNA | BC170942.1 | - | 4 - 5 | 1062 | 123 | |
BC170944.1 | - | 4 - 5 | 1062 | 123 | ||
CR761045.2 | - | 4 - 5 | 1105 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |