Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpepe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016448.2 | NP_001016448.1 | 5 - 6 | 454 | 94 | |
XM_012970425.2 | XP_012825879.1 | 5 - 6 | 506 | 94 | ||
XM_012970426.2 | XP_012825880.1 | 5 - 6 | 516 | 94 | ||
XM_012970427.2 | XP_012825881.1 | 5 - 6 | 493 | 94 | ||
XM_018097107.1 | XP_017952596.1 | 5 - 6 | 467 | 94 | ||
GenBank mRNA | BC167317.1 | - | 5 - 6 | 504 | 94 | |
BC170743.1 | - | 5 - 6 | 432 | 94 | ||
BC170745.1 | - | 5 - 6 | 432 | 94 | ||
CR760999.2 | - | 5 - 6 | 454 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |