Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016451.2 | NP_001016451.1 | 3 - 4 | 478 | 107 | |
GenBank mRNA | BC170649.1 | - | 3 - 4 | 477 | 107 | |
BC170655.1 | - | 3 - 4 | 477 | 107 | ||
CR760990.2 | - | 3 - 4 | 478 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |