Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016457.2 | NP_001016457.1 | 5 - 6 | 474 | 68 | |
GenBank mRNA | BC096514.1 | - | 5 - 6 | 466 | 68 | |
CR760965.2 | - | 5 - 6 | 474 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |