Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016458.2 | NP_001016458.1 | 2 - 3 | 624 | 107 | |
XM_012959021.2 | XP_012814475.1 | 2 - 3 | 600 | 107 | ||
XM_012959022.2 | XP_012814476.1 | 2 - 3 | 1047 | 107 | ||
GenBank mRNA | BC121844.1 | - | 2 - 3 | 1042 | 107 | |
CR760964.2 | - | 2 - 3 | 624 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |