Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016479.2 | NP_001016479.1 | 3 - 4 | 384 | 98 | |
XM_012961095.2 | XP_012816549.1 | 4 - 5 | 473 | 98 | ||
XM_012961096.1 | XP_012816550.1 | 4 - 5 | 574 | 98 | ||
GenBank mRNA | BC135405.1 | - | 4 - 5 | 442 | 98 | |
CR760893.2 | - | 3 - 4 | 384 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |