Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016526.2 | NP_001016526.1 | 3 - 4 | 298 | 95 | |
XM_012959027.1 | XP_012814481.1 | 4 - 5 | 373 | 95 | ||
GenBank mRNA | CR926198.2 | - | 3 - 4 | 298 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |