Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016545.2 | NP_001016545.1 | 10 - 11 | 1227 | 67 | |
XM_012968421.1 | XP_012823875.1 | 10 - 11 | 1184 | 67 | ||
XM_012968422.1 | XP_012823876.1 | 10 - 11 | 1315 | 67 | ||
GenBank mRNA | CR760747.2 | - | 10 - 11 | 1227 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |