Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tstd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016548.2 | NP_001016548.1 | 5 - 6 | 879 | 94 | |
XM_012962469.2 | XP_012817923.1 | 4 - 5 | 1215 | 94 | ||
XM_012962476.2 | XP_012817930.1 | 5 - 6 | 786 | 94 | ||
GenBank mRNA | CR760738.2 | - | 5 - 6 | 879 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |