Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016582.2 | NP_001016582.1 | 1 - 2 | 100 | 74 | |
XM_012962521.2 | XP_012817975.1 | 1 - 2 | 187 | 74 | ||
GenBank mRNA | BC127350.1 | - | 1 - 2 | 164 | 74 | |
CR760626.2 | - | 1 - 2 | 100 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |