Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016585.2 | NP_001016585.1 | 3 - 4 | 325 | 66 | |
XM_012968427.1 | XP_012823881.1 | 4 - 5 | 668 | 66 | ||
XM_012968428.2 | XP_012823882.1 | 4 - 5 | 416 | 66 | ||
XM_012968429.1 | XP_012823883.1 | 4 - 5 | 692 | 66 | ||
XM_012968430.2 | XP_012823884.1 | 4 - 5 | 385 | 66 | ||
GenBank mRNA | CR760620.2 | - | 3 - 4 | 325 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |