Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: limd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016606.2 | NP_001016606.1 | 4 - 5 | 1672 | 74 | |
XM_012964707.2 | XP_012820161.1 | 5 - 6 | 1943 | 74 | ||
XM_012964708.2 | XP_012820162.1 | 5 - 6 | 1879 | 74 | ||
XM_012964709.2 | XP_012820163.1 | 5 - 6 | 1873 | 74 | ||
XM_018094655.1 | XP_017950144.1 | 5 - 6 | 1734 | 74 | ||
GenBank mRNA | BC171270.1 | - | 4 - 5 | 1585 | 74 | |
CR760537.2 | - | 4 - 5 | 1672 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |