Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016607.2 | NP_001016607.1 | 2 - 3 | 229 | 66 | |
GenBank mRNA | BC158499.1 | - | 2 - 3 | 229 | 66 | |
CR760536.2 | - | 2 - 3 | 229 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |