Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016607.2 | NP_001016607.1 | 6 - 7 | 727 | 81 | |
GenBank mRNA | BC158499.1 | - | 6 - 7 | 727 | 81 | |
CR760536.2 | - | 6 - 7 | 727 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |