Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016613.2 | NP_001016613.1 | 5 - 6 | 334 | 67 | |
XM_012959033.2 | XP_012814487.1 | 5 - 6 | 372 | 67 | ||
XM_018092088.1 | XP_017947577.1 | 6 - 7 | 420 | 67 | ||
GenBank mRNA | BC170594.1 | - | 4 - 5 | 324 | 67 | |
BC170598.1 | - | 4 - 5 | 324 | 67 | ||
CR760517.2 | - | 5 - 6 | 334 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |