Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016618.2 | NP_001016618.1 | 2 - 3 | 432 | 104 | |
GenBank mRNA | BC170580.1 | - | 2 - 3 | 413 | 104 | |
BC170582.1 | - | 2 - 3 | 413 | 104 | ||
CR760505.2 | - | 2 - 3 | 432 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |