Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016623.2 | NP_001016623.1 | 4 - 5 | 377 | 78 | |
XM_012968431.2 | XP_012823885.1 | 4 - 5 | 425 | 78 | ||
GenBank mRNA | BC167937.1 | - | 4 - 5 | 372 | 78 | |
BC170855.1 | - | 4 - 5 | 368 | 78 | ||
BC170857.1 | - | 4 - 5 | 370 | 78 | ||
CR760490.2 | - | 4 - 5 | 377 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |