Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016632.2 | NP_001016632.1 | 2 - 3 | 365 | 92 | |
GenBank mRNA | BC168415.1 | - | 2 - 3 | 311 | 92 | |
BC170617.1 | - | 2 - 3 | 190 | 92 | ||
BC170619.1 | - | 2 - 3 | 188 | 92 | ||
CR760445.2 | - | 2 - 3 | 365 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |