Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016632.2 | NP_001016632.1 | 3 - 4 | 467 | 83 | |
GenBank mRNA | BC168415.1 | - | 3 - 4 | 413 | 83 | |
BC170617.1 | - | 3 - 4 | 292 | 83 | ||
BC170619.1 | - | 3 - 4 | 290 | 83 | ||
CR760445.2 | - | 3 - 4 | 467 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |