Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tekt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016652.2 | NP_001016652.1 | 3 - 4 | 418 | 89 | |
GenBank mRNA | BC118772.1 | - | 3 - 4 | 442 | 89 | |
CR760343.2 | - | 3 - 4 | 418 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |