Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zc4h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016675.2 | NP_001016675.1 | 1 - 2 | 95 | 70 | |
GenBank mRNA | BC170701.1 | - | 1 - 2 | 84 | 70 | |
BC171113.1 | - | 1 - 2 | 84 | 70 | ||
CR760233.2 | - | 1 - 2 | 95 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |