Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zc4h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016675.2 | NP_001016675.1 | 3 - 4 | 441 | 69 | |
GenBank mRNA | BC170701.1 | - | 3 - 4 | 430 | 69 | |
BC171113.1 | - | 3 - 4 | 430 | 69 | ||
CR760233.2 | - | 3 - 4 | 441 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |