Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ybx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016677.2 | NP_001016677.1 | 3 - 4 | 354 | 83 | |
GenBank mRNA | BC170833.1 | - | 3 - 4 | 338 | 83 | |
BC170835.1 | - | 3 - 4 | 338 | 83 | ||
CR760096.2 | - | 3 - 4 | 209 | 83 | ||
CR760222.2 | - | 3 - 4 | 354 | 83 | ||
CU075424.1 | - | 3 - 4 | 366 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |