Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016715.2 | NP_001016715.1 | 7 - 8 | 898 | 69 | |
GenBank mRNA | BC087776.1 | - | 7 - 8 | 909 | 69 | |
BC123027.1 | - | 7 - 8 | 908 | 69 | ||
CR760097.2 | - | 7 - 8 | 898 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |