Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016717.2 | NP_001016717.1 | 8 - 9 | 904 | 97 | |
XM_012962749.1 | XP_012818203.1 | 8 - 9 | 1038 | 97 | ||
GenBank mRNA | BC075613.1 | - | 8 - 9 | 1043 | 97 | |
BC135894.1 | - | 8 - 9 | 913 | 97 | ||
CR760093.2 | - | 8 - 9 | 904 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |