Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc39a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016721.2 | NP_001016721.1 | 4 - 5 | 462 | 69 | |
XM_018097746.1 | XP_017953235.1 | 4 - 5 | 387 | 69 | ||
GenBank mRNA | CR760068.2 | - | 4 - 5 | 462 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Xenopus tropicalis Probe spans exons |