Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsph6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016760.2 | NP_001016760.1 | 1 - 2 | 160 | 68 | |
XM_012968440.1 | XP_012823894.1 | 3 - 4 | 249 | 68 | ||
GenBank mRNA | BC127351.1 | - | 1 - 2 | 127 | 68 | |
CR855771.2 | - | 1 - 2 | 160 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |