Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsph6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016760.2 | NP_001016760.1 | 5 - 6 | 1288 | 70 | |
XM_012968440.1 | XP_012823894.1 | 7 - 8 | 1377 | 70 | ||
GenBank mRNA | BC127351.1 | - | 5 - 6 | 1255 | 70 | |
CR855771.2 | - | 5 - 6 | 1288 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |