Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cript | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016794.2 | NP_001016794.1 | 2 - 3 | 77 | 95 | |
GenBank mRNA | BC167119.1 | - | 2 - 3 | 137 | 95 | |
CR855641.2 | - | 2 - 3 | 77 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |