Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhdc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016816.2 | NP_001016816.1 | 6 - 7 | 829 | 75 | |
GenBank mRNA | CR855555.2 | - | 6 - 7 | 829 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |