Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: herpud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016818.2 | NP_001016818.1 | 2 - 3 | 463 | 80 | |
XM_012964732.2 | XP_012820186.1 | 3 - 4 | 727 | 80 | ||
XM_012964733.1 | XP_012820187.1 | 3 - 4 | 590 | 80 | ||
XM_012964734.2 | XP_012820188.1 | 3 - 4 | 727 | 80 | ||
GenBank mRNA | BC127353.1 | - | 3 - 4 | 538 | 80 | |
CR855549.2 | - | 2 - 3 | 463 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |