Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016888.2 | NP_001016888.1 | 5 - 6 | 483 | 77 | |
GenBank mRNA | BC152032.1 | - | 5 - 6 | 450 | 77 | |
CR848569.2 | - | 5 - 6 | 483 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |