Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016889.2 | NP_001016889.1 | 1 - 2 | 182 | 80 | |
GenBank mRNA | CR848568.2 | - | 1 - 2 | 182 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |