Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016889.2 | NP_001016889.1 | 6 - 7 | 1094 | 86 | |
XM_012964753.2 | XP_012820207.1 | 6 - 7 | 1332 | 86 | ||
XM_012964754.2 | XP_012820208.1 | 7 - 8 | 1492 | 86 | ||
XM_012964755.2 | XP_012820209.1 | 7 - 8 | 1700 | 86 | ||
XM_018094665.1 | XP_017950154.1 | 7 - 8 | 1189 | 86 | ||
XM_018094666.1 | XP_017950155.1 | 6 - 7 | 4117 | 86 | ||
GenBank mRNA | BC091064.1 | - | 6 - 7 | 1074 | 86 | |
CR848568.2 | - | 6 - 7 | 1094 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |